siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008725.3 | NP_001008725.2 | 4 | 525 | ||
| XM_006231928.2 | XP_006231990.1 | 6 | 778 | |||
| XM_008760736.1 | XP_008758958.1 | 6 | 1623 | |||
| GenBank mRNA | AY310138.1 | - | 495 | |||
| JQ013730.1 | - | 4 | 876 | |||
| M92340.1 | - | 4 | 559 | |||

Il6st
Genomic Map