siRNA Details
| Gene Symbol: Sdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013026.2 | NP_037158.1 | 3 | 776 | ||
| GenBank mRNA | BC061834.1 | - | 3 | 776 | 5600086 | |
| M81785.1 | - | 3 | 791 | |||
| S61865.1 | - | 3 | 791 | |||
| X60651.1 | - | 2 | 466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdc1| Gene Symbol: Sdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013026.2 | NP_037158.1 | 3 | 776 | ||
| GenBank mRNA | BC061834.1 | - | 3 | 776 | 5600086 | |
| M81785.1 | - | 3 | 791 | |||
| S61865.1 | - | 3 | 791 | |||
| X60651.1 | - | 2 | 466 | |||