siRNA Details
| Gene Symbol: Sry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012772.1 | NP_036904.1 | 1 | 309 | ||
| Gene Symbol: LOC103694552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773686.1 | XP_008771908.1 | 1 | 1391 | ||

Sry
Genomic Map