siRNA Details
| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033443.1 | NP_254278.1 | 2 | 564 | ||
| GenBank mRNA | BC127450.1 | - | 2 | 455 | ||
| BC168241.1 | - | 535 | 7934677 | |||
| CK482953.1 | - | 2 | 564 | |||
| D49434.1 | - | 2 | 306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arsb| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033443.1 | NP_254278.1 | 2 | 564 | ||
| GenBank mRNA | BC127450.1 | - | 2 | 455 | ||
| BC168241.1 | - | 535 | 7934677 | |||
| CK482953.1 | - | 2 | 564 | |||
| D49434.1 | - | 2 | 306 | |||