siRNA Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017092.1 | NP_058788.1 | 12 | 1499 | ||
| GenBank mRNA | D37880.1 | - | 12 | 1499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tyro3| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017092.1 | NP_058788.1 | 12 | 1499 | ||
| GenBank mRNA | D37880.1 | - | 12 | 1499 | ||