siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017094.1 | NP_058790.1 | 6, 7 | 809 | ||
| XM_008760752.1 | XP_008758974.1 | 6, 7 | 863 | |||
| XM_008760753.1 | XP_008758975.1 | 6, 7 | 744 | |||
| XM_008760754.1 | XP_008758976.1 | 6, 7 | 1087 | |||
| GenBank mRNA | BC086591.1 | - | 6, 7 | 792 | ||
| BC103659.1 | - | 6, 7 | 771 | |||
| BC127501.1 | - | 6, 7 | 850 | |||
| BC161852.1 | - | 6, 7 | 796 | |||
| FQ216498.1 | - | 6, 7 | 865 | |||
| FQ219220.1 | - | 6, 7 | 841 | |||
| J04811.1 | - | 6, 7 | 809 | |||
| S49003.1 | - | 6, 7 | 809 | |||

Ghr
Genomic Map