siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017094.1 | NP_058790.1 | 10 | 1245 | ||
| XM_008760752.1 | XP_008758974.1 | 10 | 1299 | |||
| XM_008760753.1 | XP_008758975.1 | 10 | 1180 | |||
| XM_008760754.1 | XP_008758976.1 | 10 | 1523 | |||
| GenBank mRNA | J04811.1 | - | 10 | 1245 | ||
| JF412704.1 | - | 7 | 707 | |||

Ghr
Genomic Map