siRNA Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012778.1 | NP_036910.1 | 4 | 769 | ||
| GenBank mRNA | BC090068.1 | - | 4 | 751 | 7190406 | |
| L07268.1 | - | 4 | 760 | |||
| X67948.1 | - | 4 | 769 | |||
| X70257.1 | - | 4 | 709 | |||
| X71069.1 | - | 4 | 720 | |||

Aqp1
Genomic Map