siRNA Details
| Gene Symbol: Aqp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012779.1 | NP_036911.1 | 1 | 239 | ||
| GenBank mRNA | BC078904.1 | - | 1 | 233 | ||
| BC092572.1 | - | 1 | 228 | 7122974 | ||
| BC110045.1 | - | 1 | 244 | 7100092 | ||
| U16245.1 | - | 1 | 239 | |||

Aqp5
Genomic Map