siRNA Details
| Gene Symbol: Bckdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012782.1 | NP_036914.1 | 8 | 1087 | ||
| GenBank mRNA | BC089915.1 | - | 6 | 734 | ||
| BC161819.1 | - | 8 | 1092 | 9099948 | ||
| J02827.1 | - | 8 | 1071 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bckdha| Gene Symbol: Bckdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012782.1 | NP_036914.1 | 8 | 1087 | ||
| GenBank mRNA | BC089915.1 | - | 6 | 734 | ||
| BC161819.1 | - | 8 | 1092 | 9099948 | ||
| J02827.1 | - | 8 | 1071 | |||