siRNA Details
| Gene Symbol: Bdkrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270713.1 | NP_001257642.1 | 4 | 427 | ||
| NM_173100.2 | NP_775123.2 | 3 | 253 | |||
| GenBank mRNA | M59967.1 | - | 4 | 421 | ||
| X69681.1 | - | 3 | 253 | |||
More Information
Set Membership: |
|

Bdkrb2
Genomic Map