siRNA Details
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012785.3 | NP_036917.2 | 3 | 361 | ||
XM_006230108.2 | XP_006230170.1 | 3 | 435 | |||
GenBank mRNA | FQ225865.1 | - | 361 | |||
U46149.1 | - | 2 | 187 |
