siRNA Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012785.3 | NP_036917.2 | 6 | 651 | ||
| XM_006230108.2 | XP_006230170.1 | 6 | 725 | |||
| GenBank mRNA | FQ225865.1 | - | 651 | |||
| U46149.1 | - | 5 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq7| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012785.3 | NP_036917.2 | 6 | 651 | ||
| XM_006230108.2 | XP_006230170.1 | 6 | 725 | |||
| GenBank mRNA | FQ225865.1 | - | 651 | |||
| U46149.1 | - | 5 | 477 | |||