siRNA Details
| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012790.2 | NP_036922.2 | 2, 3 | 159 | ||
| GenBank mRNA | AF247187.1 | - | 2, 3 | 159 | ||
| AF250373.1 | - | 2, 3 | 150 | |||
| AF250374.1 | - | 159 | ||||
| AJ403971.1 | - | 2, 3 | 159 | |||
| U02074.1 | - | 2, 3 | 162 | |||

Dspp
Genomic Map