siRNA Details
| Gene Symbol: SLC2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000340.1 | NP_000331.1 | 5 | 910 | ||
| NM_001278658.1 | NP_001265587.1 | 4 | 647 | |||
| NM_001278659.1 | NP_001265588.1 | 4 | 785 | |||
| XM_011513087.2 | XP_011511389.1 | 6 | 657 | |||
| XM_011513089.2 | XP_011511391.1 | 2 | 198 | |||
| GenBank mRNA | AK290846.1 | - | 5 | 653 | ||
| AK292741.1 | - | 5 | 653 | |||
| AK298418.1 | - | 4 | 411 | |||
| AK313622.1 | - | 5 | 651 | |||
| BC060041.1 | - | 4 | 530 | 4716156 | ||
| J03810.1 | - | 5 | 639 | |||

SLC2A2
Genomic Map