siRNA Details
| Gene Symbol: Gp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012795.2 | NP_036927.1 | 2 | 451 | ||
| XM_006248483.2 | XP_006248545.1 | 2 | 737 | |||
| Gene Symbol: LOC100911551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751076.3 | XP_003751124.2 | 1 | 449 | ||

Gp5
Genomic Map