siRNA Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012801.1 | NP_036933.1 | 5 | 666 | ||
XM_006248918.2 | XP_006248980.1 | 3 | 648 | |||
XM_006248919.2 | XP_006248981.1 | 3 | 648 | |||
XM_008768940.1 | XP_008767162.1 | 4 | 2247 | |||
GenBank mRNA | BC061731.1 | - | 3 | 653 | 5598667 | |
D10106.1 | - | 3 | 225 | |||
L06238.1 | - | 5 | 666 | |||
L06894.1 | - | 4 | 310 | |||
Z14120.1 | - | 3 | 160 |
