siRNA Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012802.1 | NP_036934.1 | 21 | 2978 | ||
| XM_006250879.2 | XP_006250941.1 | 21 | 3033 | |||
| XM_006250880.2 | XP_006250942.1 | 22 | 3339 | |||
| XM_006250881.2 | XP_006250943.1 | 21 | 2987 | |||
| GenBank mRNA | CB801421.1 | - | 2 | 152 | ||
| M63837.1 | - | 21 | 2983 | |||

Pdgfra
Genomic Map