siRNA Details
| Gene Symbol: Proc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012803.1 | NP_036935.1 | 7 | 692 | ||
| XM_006254526.2 | XP_006254588.1 | 7 | 764 | |||
| XM_006254527.2 | XP_006254589.1 | 7 | 771 | |||
| XM_008772018.1 | XP_008770240.1 | 7 | 834 | |||
| GenBank mRNA | BC078879.1 | - | 6 | 669 | 7122006 | |
| X64336.1 | - | 7 | 692 | |||

Proc
Genomic Map