siRNA Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012804.1 | NP_036936.1 | 7 | 568 | ||
| GenBank mRNA | D90038.1 | - | 7 | 568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd3| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012804.1 | NP_036936.1 | 7 | 568 | ||
| GenBank mRNA | D90038.1 | - | 7 | 568 | ||