siRNA Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270556.1 | NP_001257485.1 | 4 | 687 | ||
| NM_012806.2 | NP_036938.2 | 4 | 714 | |||
| XM_008769992.1 | XP_008768214.1 | 3 | 3506 | |||
| GenBank mRNA | DQ377224.1 | - | 1 | 61 | ||
| FQ211777.1 | - | 4 | 686 | |||
| L27128.1 | - | 4 | 424 | |||

Mapk10
Genomic Map