siRNA Details
Gene Symbol: Tst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012808.1 | NP_036940.1 | 2 | 865 | ||
GenBank mRNA | BC088449.1 | - | 2 | 865 | 7126617 | |
X56228.1 | - | 2 | 828 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012808.1 | NP_036940.1 | 2 | 865 | ||
GenBank mRNA | BC088449.1 | - | 2 | 865 | 7126617 | |
X56228.1 | - | 2 | 828 |