siRNA Details
| Gene Symbol: Cnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012809.2 | NP_036941.1 | 4 | 981 | ||
| XM_006247260.2 | XP_006247322.1 | 4 | 1001 | |||
| XM_006247261.2 | XP_006247323.1 | 4 | 1005 | |||
| XM_008767948.1 | XP_008766170.1 | 4 | 915 | |||
| GenBank mRNA | BC098066.1 | - | 4 | 981 | 7458826 | |
| L16532.1 | - | 4 | 980 | |||
| M18630.1 | - | 3 | 1274 | |||

Cnp
Genomic Map