siRNA Details
| Gene Symbol: Cyp24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201635.2 | NP_963966.1 | 10, 11 | 1441 | ||
| XM_006235672.2 | XP_006235734.1 | 10, 11 | 1783 | |||
| GenBank mRNA | BC100059.1 | - | 10, 11 | 1786 | 7129117 | |
| X59506.1 | - | 10, 11 | 1784 | |||

Cyp24a1
Genomic Map