siRNA Details
| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012813.2 | NP_036945.2 | 4 | 829 | ||
| XM_008763368.1 | XP_008761590.1 | 4 | 829 | |||
| GenBank mRNA | D45255.1 | - | 4 | 580 | ||
| U53883.1 | - | 4 | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia1| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012813.2 | NP_036945.2 | 4 | 829 | ||
| XM_008763368.1 | XP_008761590.1 | 4 | 829 | |||
| GenBank mRNA | D45255.1 | - | 4 | 580 | ||
| U53883.1 | - | 4 | 550 | |||