siRNA Details
| Gene Symbol: Amacr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012816.2 | NP_036948.2 | 5 | 1111 | ||
| GenBank mRNA | AY325250.1 | - | 1360 | |||
| BC078885.1 | - | 5 | 1079 | 7110324 | ||
| BC091163.1 | - | 5 | 1080 | 7130642 | ||
| BC127505.1 | - | 5 | 1100 | 7325347 | ||
| U89905.1 | - | 5 | 1099 | |||

Amacr
Genomic Map