siRNA Details
| Gene Symbol: Igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012817.1 | NP_036949.1 | 1 | 873 | ||
| GenBank mRNA | BC087030.1 | - | 1075 | |||
| M62781.1 | - | 1 | 873 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp5| Gene Symbol: Igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012817.1 | NP_036949.1 | 1 | 873 | ||
| GenBank mRNA | BC087030.1 | - | 1075 | |||
| M62781.1 | - | 1 | 873 | |||