siRNA Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012820.1 | NP_036952.1 | 6 | 677 | ||
| XM_006253122.1 | XP_006253184.1 | 7 | 1337 | |||
| XM_006253123.1 | XP_006253185.1 | 7 | 878 | |||
| XM_006253124.2 | XP_006253186.1 | 7 | 797 | |||
| XM_006253125.2 | XP_006253187.1 | 7 | 957 | |||
| XM_006253126.1 | XP_006253188.1 | 7 | 803 | |||
| XM_006253127.1 | XP_006253189.1 | 7 | 768 | |||
| XM_008771254.1 | XP_008769476.1 | 6 | 703 | |||
| GenBank mRNA | D90109.1 | - | 6 | 677 | ||
| FQ217275.1 | - | 6 | 647 | |||

Acsl1
Genomic Map