siRNA Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012828.2 | NP_036960.1 | 8 | 692 | ||
| XM_006257329.2 | XP_006257391.1 | 8 | 764 | |||
| XM_006257330.2 | XP_006257392.1 | 8 | 1173 | |||
| GenBank mRNA | M88751.1 | - | 8 | 696 | ||

Cacnb3
Genomic Map