siRNA Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012838.2 | NP_036970.1 | 3 | 346 | ||
| GenBank mRNA | BC084725.1 | - | 3 | 334 | 7318993 | |
| FQ220397.1 | - | 3 | 330 | |||
| FQ221069.1 | - | 3 | 306 | |||
| FQ222613.1 | - | 3 | 301 | |||
| X54737.1 | - | 3 | 338 | |||

Cstb
Genomic Map