siRNA Details
| Gene Symbol: Cycs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012839.2 | NP_036971.1 | 3 | 920 | ||
| GenBank mRNA | BC081849.1 | - | 3 | 920 | 7192255 | |
| BC089051.1 | - | 3 | 931 | 7305562 | ||
| FQ211631.1 | - | 3 | 912 | |||
| FQ212556.1 | - | 3 | 914 | |||
| FQ213464.1 | - | 3 | 908 | |||
| FQ214646.1 | - | 3 | 943 | |||
| FQ214733.1 | - | 3 | 912 | |||
| FQ214738.1 | - | 3 | 910 | |||
| FQ214838.1 | - | 3 | 911 | |||
| FQ214908.1 | - | 3 | 912 | |||
| FQ215052.1 | - | 3 | 912 | |||
| FQ215109.1 | - | 3 | 912 | |||
| FQ215466.1 | - | 3 | 912 | |||
| FQ215858.1 | - | 3 | 955 | |||
| FQ216163.1 | - | 3 | 914 | |||
| FQ216217.1 | - | 3 | 910 | |||
| FQ216281.1 | - | 3 | 970 | |||
| FQ216426.1 | - | 3 | 913 | |||
| FQ216512.1 | - | 3 | 912 | |||
| FQ216560.1 | - | 3 | 913 | |||
| FQ216674.1 | - | 3 | 911 | |||
| FQ217283.1 | - | 3 | 912 | |||
| FQ217634.1 | - | 3 | 912 | |||
| FQ218428.1 | - | 3 | 912 | |||
| FQ218668.1 | - | 3 | 913 | |||
| FQ220222.1 | - | 3 | 943 | |||
| FQ220855.1 | - | 3 | 955 | |||
| FQ223761.1 | - | 3 | 912 | |||
| FQ224710.1 | - | 3 | 912 | |||
| FQ224914.1 | - | 3 | 955 | |||
| Gene Symbol: LOC100363502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751799.3 | XP_003751847.1 | 1 | 916 | ||

Cycs
Genomic Map