siRNA Details
| Gene Symbol: Htr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012852.1 | NP_036984.1 | 1 | 839 | ||
| XM_008764208.1 | XP_008762430.1 | 2 | 1632 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr1d| Gene Symbol: Htr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012852.1 | NP_036984.1 | 1 | 839 | ||
| XM_008764208.1 | XP_008762430.1 | 2 | 1632 | |||
Set Membership: |
|