siRNA Details
| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012855.2 | NP_036987.2 | 12 | 1710 | ||
| XM_008771064.1 | XP_008769286.1 | 12 | 2808 | |||
| GenBank mRNA | D28508.1 | - | 12 | 1710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jak3| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012855.2 | NP_036987.2 | 12 | 1710 | ||
| XM_008771064.1 | XP_008769286.1 | 12 | 2808 | |||
| GenBank mRNA | D28508.1 | - | 12 | 1710 | ||