siRNA Details
Gene Symbol: Lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012857.2 | NP_036989.1 | 3 | 385 | ||
GenBank mRNA | M34959.1 | - | 3 | 385 | ||
X14765.1 | - | 2 | 221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012857.2 | NP_036989.1 | 3 | 385 | ||
GenBank mRNA | M34959.1 | - | 3 | 385 | ||
X14765.1 | - | 2 | 221 |