siRNA Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012859.1 | NP_036991.1 | 8 | 2571 | ||
| XM_006228391.2 | XP_006228453.1 | 8 | 1704 | |||
| XM_006228393.2 | XP_006228455.1 | 8 | 1955 | |||
| XM_006228394.2 | XP_006228456.1 | 8 | 3282 | |||
| XM_008758895.1 | XP_008757117.1 | 8 | 1688 | |||
| XM_008758896.1 | XP_008757118.1 | 8 | 1848 | |||
| GenBank mRNA | U40001.1 | - | 8 | 2571 | ||
| X51415.1 | - | 7 | 2093 | |||

Lipe
Genomic Map