siRNA Details
| Gene Symbol: Mgmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012861.1 | NP_036993.1 | 4 | 416 | ||
| GenBank mRNA | M76704.1 | - | 3 | 361 | ||
| S61804.1 | - | 4 | 416 | |||
| X54862.1 | - | 4 | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgmt| Gene Symbol: Mgmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012861.1 | NP_036993.1 | 4 | 416 | ||
| GenBank mRNA | M76704.1 | - | 3 | 361 | ||
| S61804.1 | - | 4 | 416 | |||
| X54862.1 | - | 4 | 442 | |||