siRNA Details
| Gene Symbol: Ninj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012867.1 | NP_036999.1 | 2 | 264 | ||
| GenBank mRNA | FQ215609.1 | - | 2 | 296 | ||
| FQ219073.1 | - | 2 | 296 | |||
| FQ220812.1 | - | 2 | 296 | |||
| FQ225610.1 | - | 2 | 268 | |||
| FQ229388.1 | - | 2 | 296 | |||
| FQ230574.1 | - | 2 | 268 | |||
| FQ232057.1 | - | 2 | 296 | |||
| U72660.1 | - | 2 | 286 | |||

Ninj1
Genomic Map