siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 2, 3 | 885 | ||
| XM_006232045.2 | XP_006232107.1 | 2, 3 | 1469 | |||
| GenBank mRNA | L27339.1 | - | 2, 3 | 885 | ||
| X78595.1 | - | 731 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npr3| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 2, 3 | 885 | ||
| XM_006232045.2 | XP_006232107.1 | 2, 3 | 1469 | |||
| GenBank mRNA | L27339.1 | - | 2, 3 | 885 | ||
| X78595.1 | - | 731 | ||||