siRNA Details
| Gene Symbol: Npy5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012869.1 | NP_037001.1 | 2 | 385 | ||
| XM_006253013.2 | XP_006253075.1 | 2 | 240 | |||
| XM_006253014.2 | XP_006253076.1 | 3 | 430 | |||
| GenBank mRNA | AF044264.1 | - | 3 | 443 | ||
| U56078.1 | - | 1 | 202 | |||
| U66274.1 | - | 2 | 385 | |||
More Information
Set Membership: |
|

Npy5r
Genomic Map