siRNA Details
| Gene Symbol: Oxtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012871.2 | NP_037003.2 | 1 | 628 | ||
| XM_006237008.2 | XP_006237070.1 | 2 | 1753 | |||
| XM_008763179.1 | XP_008761401.1 | 2 | 2062 | |||
| XM_008763180.1 | XP_008761402.1 | 2 | 1850 | |||
| XM_008763181.1 | XP_008761403.1 | 2 | 2043 | |||
| XM_008763182.1 | XP_008761404.1 | 2 | 1557 | |||
| GenBank mRNA | AF257237.1 | - | 1 | 820 | ||
| AY338194.1 | - | 1 | 628 | |||
| L81169.1 | - | 1 | 658 | |||
More Information
Other Species Targeted
Set Membership: |
|

Oxtr
Genomic Map