siRNA Details
| Gene Symbol: Pdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012872.1 | NP_037004.1 | 4 | 649 | ||
| GenBank mRNA | M33528.1 | - | 4 | 649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdc| Gene Symbol: Pdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012872.1 | NP_037004.1 | 4 | 649 | ||
| GenBank mRNA | M33528.1 | - | 4 | 649 | ||