siRNA Details
| Gene Symbol: Phb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031851.2 | NP_114039.1 | 7 | 1028 | ||
| GenBank mRNA | BC072518.1 | - | 7 | 1028 | 7105633 | |
| FQ229103.1 | - | 7 | 1030 | |||
| M61219.1 | - | 6 | 980 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phb| Gene Symbol: Phb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031851.2 | NP_114039.1 | 7 | 1028 | ||
| GenBank mRNA | BC072518.1 | - | 7 | 1028 | 7105633 | |
| FQ229103.1 | - | 7 | 1030 | |||
| M61219.1 | - | 6 | 980 | |||