siRNA Details
| Gene Symbol: Ros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012874.1 | NP_037006.1 | 15, 16 | 2519 | ||
| XM_008772862.1 | XP_008771084.1 | 15, 16 | 2637 | |||
| XM_008772863.1 | XP_008771085.1 | 19, 20 | 2365 | |||
| GenBank mRNA | M35104.1 | - | 15, 16 | 2456 | ||
| M35105.1 | - | 15, 16 | 2456 | |||
| M35106.1 | - | 15, 16 | 2519 | |||

Ros1
Genomic Map