siRNA Details
Gene Symbol: LOC103692785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770106.1 | XP_008768328.1 | 1 | 174 |
Gene Symbol: LOC103690821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773208.1 | XP_008771430.1 | 1 | 140 |
Gene Symbol: LOC103692831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765320.1 | XP_008763542.1 | 1 | 170 |
Gene Symbol: LOC103693375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770613.1 | XP_008768835.1 | 1 | 173 |
Gene Symbol: LOC103694169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771991.1 | XP_008770213.1 | 1 | 170 |
Gene Symbol: LOC103694404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772938.1 | XP_008771160.1 | 1 | 175 |
Gene Symbol: Rpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012875.2 | NP_037007.1 | 140 | |||
GenBank mRNA | BC058489.1 | - | 174 | 6887843 | ||
X82551.1 | - | 140 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
