siRNA Details
| Gene Symbol: Scn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012877.1 | NP_037009.1 | 2 | 338 | ||
| XM_006242895.2 | XP_006242957.1 | 2 | 290 | |||
| GenBank mRNA | U37026.1 | - | 2 | 338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn2b| Gene Symbol: Scn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012877.1 | NP_037009.1 | 2 | 338 | ||
| XM_006242895.2 | XP_006242957.1 | 2 | 290 | |||
| GenBank mRNA | U37026.1 | - | 2 | 338 | ||