siRNA Details
| Gene Symbol: Slc2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012879.2 | NP_037011.2 | 7 | 914 | ||
| XM_006232207.2 | XP_006232269.1 | 8 | 1218 | |||
| GenBank mRNA | BC078875.1 | - | 7 | 914 | 7127751 | |
| J03145.1 | - | 7 | 989 | |||

Slc2a2
Genomic Map