siRNA Details
| Gene Symbol: Sstr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012882.2 | NP_037014.2 | 2 | 361 | ||
| GenBank mRNA | L04535.1 | - | 2 | 361 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sstr5| Gene Symbol: Sstr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012882.2 | NP_037014.2 | 2 | 361 | ||
| GenBank mRNA | L04535.1 | - | 2 | 361 | ||
Set Membership: |
|