siRNA Details
| Gene Symbol: Cntn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012884.1 | NP_037016.1 | 8 | 1121 | ||
| XM_006249746.2 | XP_006249808.1 | 9 | 1335 | |||
| XM_008769427.1 | XP_008767649.1 | 8 | 1224 | |||
| GenBank mRNA | M31725.1 | - | 8 | 1121 | ||
More Information
Set Membership: |
|

Cntn2
Genomic Map