siRNA Details
| Gene Symbol: Vcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012889.1 | NP_037021.1 | 248 | |||
| GenBank mRNA | BC089812.1 | - | 266 | 7364929 | ||
| M84488.1 | - | 248 | ||||
| X63722.1 | - | 197 | ||||
| Gene Symbol: LOC100912479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749384.3 | XP_003749432.1 | 2 | 461 | ||

Vcam1
Genomic Map