siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012891.2 | NP_037023.1 | 10, 11 | 1103 | ||
| GenBank mRNA | BC087036.1 | - | 10, 11 | 1124 | 7193535 | |
| D30647.1 | - | 10, 11 | 1105 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acadvl| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012891.2 | NP_037023.1 | 10, 11 | 1103 | ||
| GenBank mRNA | BC087036.1 | - | 10, 11 | 1124 | 7193535 | |
| D30647.1 | - | 10, 11 | 1105 | |||